Anti-NIRF Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-6389R-CY7
BOSSBS-6389R-CY7EA 893 EUR
BOSSBS-6389R-CY7
Anti-NIRF Rabbit Polyclonal Antibody (Cy7®)
Antibodies
E3 ubiquitin-protein ligase which mediates ubiquitination and subsequent proteasomal degradation of PCNP. May participate in methylation-dependent transcriptional regulation. Important for G1/S transition. Overexpression causes G1 phase cell arrest.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NIRF
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR