Anti-WDR61 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-6886R-A750
BOSSBS-6886R-A750EA 893 EUR
BOSSBS-6886R-A750
Anti-WDR61 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
WDR61 is a subunit of the human PAF and SKI complexes, which function in transcriptional regulation and are involved in events downstream of RNA synthesis, such as RNA surveillance (Zhu et al., 2005 [PubMed 16024656]).[supplied by OMIM, Mar 2008].

Type: Primary
Antigen: WDR61
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-220/305
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR