Anti-FOXN1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-6970R-A350
BOSSBS-6970R-A350EA 915 EUR
BOSSBS-6970R-A350
Anti-FOXN1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Mutations in the winged-helix transcription factor gene at the nude locus in mice and rats produce the pleiotropic phenotype of hairlessness and athymia, resulting in a severely compromised immune system. This gene is orthologous to the mouse and rat genes and encodes a similar DNA-binding transcription factor that is thought to regulate keratin gene expression. A mutation in this gene has been correlated with T-cell immunodeficiency, the skin disorder congenital alopecia, and nail dystrophy. Alternative splicing in the 5' UTR of this gene has been observed. [provided by RefSeq, Jul 2008].

More Product Information
Order Now

SPECIFICATIONS

Antigen symbol FOXN1
Environmentally Preferable
Antigen name Forkhead box protein N1
Antigen synonyms FKHL20|WHN|RONU
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 8456
Isotype IgG
ImmunoChemistry Yes
Immunogen 350-400/648
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR