Anti-TMEM263 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-6987R-A555
BOSSBS-6987R-A555EA 893 EUR
BOSSBS-6987R-A555
Anti-TMEM263 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
C12orf23 (chromosome 12 open reading frame 23), also known as FLJ11721, FLJ13959 or MGC17943, is a 116 amino acid multi-pass membrane protein belonging to the UPF0444 family. C12orf23 is encoded by a gene located on human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a number of skeletal deformities, including hypochondrogenesis, achondrogenesis and Kniest dysplasia. Noonan syndrome, which includes heart and facial developmental defects among the primary symptoms, is caused by a mutant form of PTPN11 geneMore Product Information