Anti-DNAJA3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-7645R-CY5
BOSSBS-7645R-CY5EA 900 EUR
BOSSBS-7645R-CY5
Anti-DNAJA3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
TID1 is a human homolog of the Drosophila tumor suppressor lethal tumerous imaginal discs and encodes two mitochondrial matrix localized splice variants of human Tid1 designated hTid1S and hTid1L. These proteins are the conserved members of the DnaJ family of proteins which act as cochaperons for mitochondrial Hsp70. They contain a conserved tetrahedrical J domain which binds to Hsp70 chaperones and activates their ATPase activity. Expression of hTid1L increases apoptosis induced by DNA damaging agents as mitomycin C and TNF alpha. A J domain mutant of hTid1L can dominantly suppressMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol DNAJA3
Antigen name DnaJ (Hsp40) Homolog, Subfamily A, Member 3
Environmentally Preferable
Antigen synonyms HCA57|TID1|hTID-1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 9093
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR