Anti-DPYS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Supplier: Bioss
BS-8287R-A647
BOSSBS-8287R-A647EA 893 EUR
BOSSBS-8287R-A647
Anti-DPYS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Catalyzes the second step of the reductive pyrimidine degradation, the reversible hydrolytic ring opening of dihydropyrimidines. Can catalyzes the ring opening of 5,6-dihydrouracil to N-carbamyl-alanine and of 5,6-dihydrothymine to N-carbamyl-amino isobutyrate.Tissue specificity:Liver and kidney.Involvement in disease:Defects in DPYS are the cause of dihydropyrimidinase deficiency (DHPD). DHPD is an autosomal recessive disorder characterized by dihydropyrimidinuria and associated with a variable clinical phenotype: epileptic or convulsive attacks, dysmorphic features and severe developmentalMore Product Information