Anti-FOXA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-8634R-HRP
BOSSBS-8634R-HRPEA 900 EUR
BOSSBS-8634R-HRP
Anti-FOXA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Transcription factor that is involved in embryonic development, establishment of tissue-specific gene expression and regulation of gene expression in differentiated tissues. Is thought to act as a 'pioneer' factor opening the compacted chromatin for other proteins through interactions with nucleosomal core histones and thereby replacing linker histones at target enhancer and/or promoter sites. Binds DNA with the consensus sequence 5'-[AC]A[AT]T[AG]TT[GT][AG][CT]T[CT]-3' (By similarity). Proposed to play a role in translating the epigenetic signatures into cell type-specific enhancer-drivenMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol FOXA1
Antigen name Forkhead box protein A1
Environmentally Preferable
Antigen synonyms HNF-3a|MGC33105|FoxA4a|MGC52590|pintallavis|tcf3a|fkh|xfkh2|FOXA1|Tcf3a|Tcf-3a|fkd7|forkhead-7|tFoxA1|HNF3A|HNF3-alpha|TCF3A|MGC75967|Ci-fkh|Hnf-3a|xfkh1|foxa1|HNF3alpha|Hnf3a
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 3169
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Immunogen 145-185/472
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR