Anti-GPBAR1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8874R
BOSSBS-8874REA 725 EUR
BOSSBS-8874R
Anti-GPBAR1 Rabbit Polyclonal Antibody
Antibodies
The G protein-coupled receptor TGR5 is a 330-amino acid protein that is almost universally expressed in human tissues including heart, skeletal muscle, spleen, kidney, liver, small intestine, placenta, and leukocytes, but not in brain, colon (without mucosa), thymus, or lung. TGR5 is sensitive to bile acids and responds through a significant mechanism that coordinates energy homeostasis. Bile acids activate mitogen-activated protein (MAP) kinase pathways, specifically induce TGR5 internalization, promote an increase of guanosine 5'-O-3-thio-triphosphate binding in membrane fractions, andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol GPBAR1
Antigen name G Protein-Coupled Bile Acid Receptor 1
Environmentally Preferable
Antigen synonyms M-BAR|G-protein coupled receptor 19|GPR131|TGR5|G protein-coupled bile acid receptor 1|G protein-coupled bile acid receptor|BG37|GPCR19
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 151306
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Flow cytometry Yes
Form liquid
Purification Purified by Protein A.
Concentration 1 ug/ul
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR