Anti-GPBAR1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-8874R-CY5.5
BOSSBS-8874R-CY5.5EA 900 EUR
BOSSBS-8874R-CY5.5
Anti-GPBAR1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The G protein-coupled receptor TGR5 is a 330-amino acid protein that is almost universally expressed in human tissues including heart, skeletal muscle, spleen, kidney, liver, small intestine, placenta, and leukocytes, but not in brain, colon (without mucosa), thymus, or lung. TGR5 is sensitive to bile acids and responds through a significant mechanism that coordinates energy homeostasis. Bile acids activate mitogen-activated protein (MAP) kinase pathways, specifically induce TGR5 internalization, promote an increase of guanosine 5'-O-3-thio-triphosphate binding in membrane fractions, andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol GPBAR1
Antigen name G Protein-Coupled Bile Acid Receptor 1
Environmentally Preferable
Antigen synonyms M-BAR|G-protein coupled receptor 19|GPR131|TGR5|G protein-coupled bile acid receptor 1|G protein-coupled bile acid receptor|BG37|GPCR19
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5.5®
Reactivity Human
Host Rabbit
Gene ID 151306
Isotype IgG
ImmunoChemistry Yes
Flow cytometry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR