Anti-ACCSL Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-9062R-A647
BOSSBS-9062R-A647EA 1350 EUR
BOSSBS-9062R-A647
Anti-ACCSL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Belongs to the class-I pyridoxal-phosphate-dependent aminotransferase family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACCSL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR