Anti-MEAF6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-9219R-A350
BOSSBS-9219R-A350EA 893 EUR
BOSSBS-9219R-A350
Anti-MEAF6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
MEAF6, also known as Eaf6 or NY-SAR-91, is a 191 amino acid nuclear protein belonging to the EAF6 family. MEAF6 is a component of the NuA4 histone acetyltransferase complex, which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. The gene encoding MEAF6 localizes to chromosome 1 and, due to alternative splicing events, MEAF6 exists in at least three isoforms. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol MEAF6
Antigen name MYST/Esa1-associated factor 6
Environmentally Preferable
Antigen synonyms C1orf149|EAF6|NY-SAR-91|CENP-28
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 64769
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR