Anti-LRCH3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-9329R-A680
BOSSBS-9329R-A680EA 893 EUR
BOSSBS-9329R-A680
Anti-LRCH3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Members of the leucine-rich repeat family includes LRCH1, LRCH2, LRCH3 and LRCH4. All family members contain one calponin-homology domain and nine leucine-rich repeats. The best characterised leucine-rich repeat family member is LRCH4, which is suggested to be involved in ligand binding in the brain, with expression observed primarily in the hippocampus. As a cell adhesion molecule and signal receptor, LRCH4 may play an important role in maintenance of hippocampus-dependent memories, with defects in the gene possibly contributing to a loss of long-term memory. The gene encoding LRCH3 maps toMore Product Information