Anti-PGGT1B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-9548R-A750
BOSSBS-9548R-A750EA 893 EUR
BOSSBS-9548R-A750
Anti-PGGT1B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Mammalian protein farnesyl transferases are heterodimeric proteins containing two nonidentical Alpha and beta subunits that attach farnesyl residues to a cysteine at the fourth position from the COOH terminus of several proteins, including nuclear lamins and p21Ras proteins. The natural substrates contain the Cys-A-A-Xaa recognition sequence, where the A residues are aliphatic and Xaa represents methionine, serine, glutamine or cysteine. The purified farnesyl transferase is an a-b heterodimer. The beta subunit, which is known as FT beta, CAAX farnesyltransferase subunit beta, or Ras proteinsMore Product Information