Anti-C1orf31 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-9785R-A680
BOSSBS-9785R-A680EA 893 EUR
BOSSBS-9785R-A680
Anti-C1orf31 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. C1orf31 is a 125 amino acid mitochondrial protein that belongs to the cytochrome c oxidase subunit 6B family. There are three isoforms of C1orf31 that are produced as a result of alternative splicing events. The gene encoding C1orf31 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome.More Product Information
Order Now

SPECIFICATIONS

Antigen name C1orf31
Environmentally Preferable
Antigen synonyms Uncharacterized protein C1orf31|C1orf31|RP5-827C21.3|Chromosome 1 open reading frame 31|CA031_HUMAN|Hypothetical protein LOC388753
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 388753
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 51-125/125
Immunogen KLH conjugated synthetic peptide derived from human C1orf31
Form Liquid
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR