Infusion needles with filters, Sterifix®
Supplier: B. Braun
4550404
BRAU4550404EA 91.5 EUR
BRAU4550404
Infusion needles with filters, Sterifix®
Infusions
Infusion filter needles designed for the prevention of particulate contamination.
  • Thin-walled needle (G19)
  • Particle filter and Luer fitting