Thermal transfer printer labels, type B-425, WorkHorse™
Supplier: Brady
031322 031327 031321 031328 031326 031320 030196
BRDY031322EA 137.52 EUR
BRDY031322 BRDY031327 BRDY031321 BRDY031328 BRDY031326 BRDY031320 BRDY030196
Thermal transfer printer labels, type B-425, WorkHorse™
Labels
Printable, polypropylene labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.