Labels for BBP3x printers, type B-526
Supplier: Brady
117924 117926 117928
BRDY117924EA 682.31 EUR
BRDY117924 BRDY117926 BRDY117928
Labels for BBP3x printers, type B-526
Labels
Type B-526 phosphorescent polyester tape is designed for directional and evacuation marking. It is used for glow-in-the-dark identification up to 10 hours.