Labels for BBP3x Printers, B-569
Supplier: Brady
117991 117992 117993 117990 117930 117949 117954 117962 117966 117969 117985 117987 117986 117931 117982 117965 117967 117968 117970 117971 117972 117977 117978 117979 117963 117981 117961 117983 117984 117988 117989 117980 117945 117932 117934 117936 117939 117940 117941 117942 117964 117929 117947 117948 117950 117951 117952 117956 117957 117958 117959 117943
BRDY117991EA 305.49 EUR
BRDY117991 BRDY117992 BRDY117993 BRDY117990 BRDY117930 BRDY117949 BRDY117954 BRDY117962 BRDY117966 BRDY117969 BRDY117985 BRDY117987 BRDY117986 BRDY117931 BRDY117982 BRDY117965 BRDY117967 BRDY117968 BRDY117970 BRDY117971 BRDY117972 BRDY117977 BRDY117978 BRDY117979 BRDY117963 BRDY117981 BRDY117961 BRDY117983 BRDY117984 BRDY117988 BRDY117989 BRDY117980 BRDY117945 BRDY117932 BRDY117934 BRDY117936 BRDY117939 BRDY117940 BRDY117941 BRDY117942 BRDY117964 BRDY117929 BRDY117947 BRDY117948 BRDY117950 BRDY117951 BRDY117952 BRDY117956 BRDY117957 BRDY117958 BRDY117959 BRDY117943
Labels for BBP3x Printers, B-569
Labels
This high performance polyester labeling tape is used for labeling stainless steel pipes and surfaces. Applies best to very smooth surfaces and ideal for mounting to rigid panels.