Repositionable vinyl labels for BMP21-PLUS, BMP21-LAB, M210, M210-LAB, and M211 label printers
Supplier: Brady
152246 152247
BRDY152246EA 33.47 EUR
BRDY152246 BRDY152247
Repositionable vinyl labels for BMP21-PLUS, BMP21-LAB, M210, M210-LAB, and M211 label printers
Labels
Repositionable vinyl film (B-581) is a durable, flexible, high-gloss material ideal for indoor/outdoor marking where frequent repositioning is required.
  • Ideal for applications such as 5S and inventory
  • Vinyl film can be thermal transfer printed
  • Average outdoor durability is 5 years
  • Service temperature: −40 to 82 °C
Special aggressive adhesive stays in place but removes cleanly without leaving adhesive residue.