Aggressive Adhesive Self-Laminating Vinyl Wrap Around Labels for M710 Printer
Supplier: Brady
170485 170489 170491 170490 170484 170483 170487 170488 170492 170486
BRDY170485EA 55.78 EUR
BRDY170485 BRDY170489 BRDY170491 BRDY170490 BRDY170484 BRDY170483 BRDY170487 BRDY170488 BRDY170492 BRDY170486
Aggressive Adhesive Self-Laminating Vinyl Wrap Around Labels for M710 Printer
Labels
B-417 High-adhesion self-laminating vinyl wraps are designed for tough conditions, including high-humidity environments, small diameter wires and newer wire jacketing materials.
  • Dependable, long-term adhesion to small diameter wires
  • Will stay adhered in even the most humid environments
  • Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC
  • Features a white printable area and a clear overlaminating area that protects the print
  • Ideal for control panels, electrical panels, wire harnesses, datacom,