High Tensile Polypropylene Inserts/Tags with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
170724 170725 170726
BRDY170724EA 80.08 EUR
BRDY170724 BRDY170725 BRDY170726
High Tensile Polypropylene Inserts/Tags with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Labels
Mark outlet faceplates and patch panels with the B-412 high tensile strength polypropylene insert stock material. Print almost anywhere with a compact Brady portable label printer.
  • Specialised - Designed for easily marking port, jack and outlet faceplates
  • Tough - Resistant to tearing, fuels, oils, chemicals and extreme temperatures
  • Non-adhesive - Easy to place, remove and replace as needed
Application: Component and equipment labelling, data and telecommunications labelling, electrical labelling, wire and cable labelling.
Matte finish, chemical-More Product Information