Printer Ribbons, M710 R6700 Series
Supplier: Brady
173187
BRDY173187EA 96.77 EUR
BRDY173187
Printer Ribbons, M710 R6700 Series
Printer Ribbons
One-color ribbon for printing a single color onto a label.
  • R6700 series resin/wax formulation is a white ribbon that offers good chemical resistance when printed on the appropriate Brady® label material
  • Recommended for printing on dark-colored, heat shrinkable tubing materials, such as Brady® black B-342 and B-344 PermaSleeve® wire markers
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles,More Product Information