Ultra Aggressive Adhesive Multi-Purpose Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Supplier: Brady
174263 174235 174297 174303 174284 173687 173881 173817 173719 173743 173684 173730 173828
BRDY174263EA 78.5 EUR
BRDY174263 BRDY174235 BRDY174297 BRDY174303 BRDY174284 BRDY173687 BRDY173881 BRDY173817 BRDY173719 BRDY173743 BRDY173684 BRDY173730 BRDY173828
Ultra Aggressive Adhesive Multi-Purpose Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Labels
Ultra Aggressive Polyester (B-489A) with permanent adhesive is ideal for labelling products and general ID in both commercial and industrial applications.
  • Best for application on smooth and highly textured indoor surfaces, including metal and low surface energy plastics
  • Matte white polyester material has a permanent rubber based adhesive and is thermal transfer printable
  • Service temperature of −40 to 120 °C (−40 to 248 °F)
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, electricalMore Product Information