PermaSleeve® Commercial Grade Heat-Shrink Labels for M610, M611, BMP61, M710 and BMP71
Supplier: Brady
174478 174484 174464 174502 174424 174459 174477 174418 174496 174488 174471 174442 174436
BRDY174478EA 639.28 EUR
BRDY174478 BRDY174484 BRDY174464 BRDY174502 BRDY174424 BRDY174459 BRDY174477 BRDY174418 BRDY174496 BRDY174488 BRDY174471 BRDY174442 BRDY174436
PermaSleeve® Commercial Grade Heat-Shrink Labels for M610, M611, BMP61, M710 and BMP71
Labels
PermaSleeve® commercial grade wire markers provide business-class performance at a commercial price, so users can easily complete wire identification projects throughout their facilities.
  • Great for electrical contractors, maintenance crews, industrial manufacturers, and more
  • Heat-shrink sleeves provide permanent identification without any extra steps
  • Legible legends in temperatures ranging from −55 to +135 °C (−67 to +275 °F)
  • Matte finish
Applications include electrical labelling and wire and cable labelling.
Brady authentic materials workMore Product Information