Engraved Replcement Labels for M710, M610, M611
Supplier: Brady
312102 312094 312120 312110 312071 312111 312116 312073 312106 312096 312113 312119 312112 312069 312125 312104 312114 312124 312093 312070 312092 312072 312118 312103 312121 312095 312117 312068 312105 312091
BRDY312102EA 71.61 EUR
BRDY312102 BRDY312094 BRDY312120 BRDY312110 BRDY312071 BRDY312111 BRDY312116 BRDY312073 BRDY312106 BRDY312096 BRDY312113 BRDY312119 BRDY312112 BRDY312069 BRDY312125 BRDY312104 BRDY312114 BRDY312124 BRDY312093 BRDY312070 BRDY312092 BRDY312072 BRDY312118 BRDY312103 BRDY312121 BRDY312095 BRDY312117 BRDY312068 BRDY312105 BRDY312091
Engraved Replcement Labels for M710, M610, M611
Labels
Engraved plate replacement labels for the M610 and M611 thermal transfer portable printers. These parts can also be printed on the M710 label printer provided the roll adapter (M71-ADAPT) is used.
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
  • Polyethylene foam laminate Polyester material
  • Service temperature of −40 to +100 °C
  • Gloss finish
Ideal for data and telecommunications labelling, electrical labelling, and general identification applications.