Magicard™ Monochrome Film, Silver
Supplier: Brady
321965
BRDY321965EA 75 EUR
BRDY321965
Magicard™ Monochrome Film, Silver
Printer Ribbons
The Magicard™ MA1000K silver ribbon is perfect for adding silver accents to prints.
  • High-quality, monochrome ribbon guarantees to print 1000 single-sided cards
  • Includes a cleaning roller for dust-free printing
Applications: Healthcare and hospital labelling, facility identification, security identification, general identification.
Delivery information: Includes 1 dye film and 1 cleaning roller.