Magicard™ Colour Film + Transfer Tape Set for Prima 8
Supplier: Brady
321977
BRDY321977EA 365 EUR
BRDY321977
Magicard™ Colour Film + Transfer Tape Set for Prima 8
Printer Ribbons
Set of dye film and transfer film all in one.
  • Print full colour on one side and black on the other
  • Print 1000 cards front and back in one go
Dye film and re-transfer film that fits the Prima 8 printers.
Delivery information: Includes 1 YMCK dye film and 1 re-transfer film.