Micro burettes, Bang pattern, straight stopcock, Schellbach stripe, BLAUBRAND®
Supplier: Brand
24265 24269 924265
BRND24265EA 381 EUR
BRND24265 BRND24269 BRND924265
Micro burettes, Bang pattern, straight stopcock, Schellbach stripe, BLAUBRAND®
Burettes
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • With base
Certifikace: DIN EN ISO 385
Informace pro objednávání: Supplied with batch certificate or single certificate. On request also available with USP individual certificate or DAkkS calibration certificate.