Repetitive pipette, HandyStep® touch and HandyStep® touch S
Supplier: Brand
705210 705200 705201 705211 705200-AT
613-6205EA 1050 EUR
613-6205 613-6204 613-6414 613-6415 BRND705200-AT
Repetitive pipette, HandyStep® touch and HandyStep® touch S
Pipettes Repetitive Pipettes
Repetitive pipettes are the first choice when it comes to efficient dispensing of long series. Positive displacement pipettes are often the right solution for pipetting viscous media. The new HandyStep touch stepper combines both features. The HandyStep touch S has additional capabilities, such as titration or sequential dispensing.
  • Touch screen operation with intuitive menu navigation in English, German, Spanish, French and Chinese
  • HandyStep® touch features multi-dispensing, auto-dispensing and pipetting; HandyStep® touch S offers the following additional
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 and ISO 50001