Anti-Gastrin Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1497-50
BSENR-1497-50EA 321 EUR
BSENR-1497-50
Anti-Gastrin Rabbit Polyclonal Antibody
Antibodies
Human Gastrin is a 101 amino acid hormone produced by G cells of the duodenum, stomach and pancreas. It stimulates secretion of hydrochloric acid by parietal cells of the stomach. Gastrin is also secreted into bloodstream.

Antibody recognizes Gastrin in multiple species and in native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should beMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol GAST
Environmentally Preferable
Antigen name Gastrin
Antigen synonyms Gastrin component III|Gastrin-34|GAST_HUMAN|Gastrin-17|Gastrin-6|Gastrin component I|Gastrin component II|Gastrin precursor
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Epitope Human Gastrin peptide (76-85 aa)
Immunogen Synthetic human Gastrin peptide (76-85 aa) conjugated to BSA.
Format Lyophilized
Cross adsorption No

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR