Disinfection systems, Phileas®
Supplier: Basan
PHI25
BSNCPHI25EA 7920 EUR
BSNCPHI25
Disinfection systems, Phileas®
Disinfection Systems
This airborne disinfection system is based on a patented technology using a rotating disk to deliver an active agent in the form of microscopic droplets in the atmosphere. The range of this system is from less than 1 m³ to several thousand m³ with various configurations.
  • Various types are available
  • Minimum maintenance
Used in the varous fields such as healthcare, pharmaceutical, agri-food, cosmetic industries, laboratories, biotechnology and animal facilities.