Needle array electrodes, AgilePulse™
Supplier: BTX
47-0050 47-0040 47-0060 47-0080 47-0070 47-0043 47-0045 47-0086
731-0329EA 362 EUR
731-0329 731-0328 731-0330 731-0332 731-0331 BTXI47-0043 BTXI47-0045 BTXI47-0086
Needle array electrodes, AgilePulse™
Electrodes Electroporation Electrodes
AgilePulse™ needle array electrodes are miniature needles designed to provide superior, highly uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ in vivo ID (intra dermal) or IM (intra muscular) gene delivery electroporation system.
  • Uniform, reliable electric fields
  • Miniature needles minimise tissue trauma
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations and sizes available
Applications for these electrodes include intra-dermal DNA vaccine research and antibodyMore Product Information