Ball valves with thread
Supplier: BURKLE
0500-8203 0500-8103 0500-8102 0500-8201 0500-8202
BURK0500-8203EA 25.5 EUR
BURK0500-8203 BURK0500-8103 BURK0500-8102 BURK0500-8201 BURK0500-8202
Ball valves with thread
Stopcocks
Ball valve made of PP with outer/inner thread.
  • Maximum operating pressure 10 bar (at +20 °C)
  • Maximum operating temperature 100 °C
  • Suitable for food
It also consists fibre-glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and a seal made of HDPE.