Eyewash bottle
Supplier: BURKLE
2507-2000
BURK2507-2000EA 43.2 EUR
BURK2507-2000
Eyewash bottle
First Aid Supplies Eyewash Stations and Eyewash Bottles
Empty bottle for 600 ml contents, leakproof due to valve and dust protection cap. Instructions printed on (in German), made of transparent PE.