Forceps, stainless steel
Supplier: BURKLE
5386-0104 5386-0304 5386-0102
BURK5386-0104EA 4.65 EUR
BURK5386-0104 BURK5386-0304 BURK5386-0102
Forceps, stainless steel
Forceps
These stainless steel forceps are suitable for precise laboratory tasks. The laboratory tweezers are made of stainless steel and feature a fluting at the ends and grip for secure handling.
  • Stainless steel (1.4301)
  • Flutings at the end and grip for secure handling
These laboratory forceps are suitable for precise work in the fields of chemistry, pharmacy, cosmetics and food and beverage.