Dosing pumps, Dosi-Pump
Supplier: BURKLE
5607-3030 5607-4100 5607-5250 5607-2030 5607-1004
BURK5607-3030EA 28.15 EUR
BURK5607-3030 181-9906 BURK5607-5250 BURK5607-2030 BURK5607-1004
Dosing pumps, Dosi-Pump
Pumps Drum/Transfer Pumps
Dosi pump are hand controlled pump used for the controlled product extraction from canisters, bottles, barrels and carboys.
  • Medium contacting parts made of PP, PE and stainless steel
  • Suitable for products of different viscosity
Dosi pump is inserted into container closures with suitable holes. Immersion value represents the value of immersion depth.