Vacuum hand pumps, VacuMan
Supplier: BURKLE
5620-1360 5620-0360 5620-2181 5620-1180 5620-2180
331-2123EA 153.85 EUR
331-2123 BURK5620-0360 BURK5620-2181 BURK5620-1180 BURK5620-2180
Vacuum hand pumps, VacuMan
Pumps Vacuum Hand Pumps
The vacuum hand pump can be operated with one hand to create a vacuum or pressure, independent of power supply.
  • Maintenance-free, self-lubricating
  • Corrosion resistant
  • With or without gauges
  • Available in PVC or aluminium for heavy duty
Connection for hoses with 6,4 mm inner diameter.
Informace o príslušenství: Sampling set with suction hose, sample container and adapter set for hoses from 5 to 14 mm available as accessory