Bunsen burners, LABOGAZ 206
Supplier: CAMPING GAZ
2000020656
CAMP2000020656EA 84 EUR
CAMP2000020656
Bunsen burners, LABOGAZ 206
Heaters Gas Burners
For use with Campingaz® pierceable cartridge C 206 GLS. Ideal use in wide range of areas such as laboratories, schools, jewelers, opticians and handicrafts.
  • Attached with clip and go installation system which makes changing cartridges easy