Anti-CB1 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
10006590-1
CAYM10006590-1EA 451 EUR
CAYM10006590-1
Anti-CB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CB1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR