Anti-PSD95 Mouse Monoclonal Antibody [clone: 7E3]

Supplier: Cayman Chemical
10011436-100 10011436-25
CAYM10011436-100EA 683 EUR
CAYM10011436-100 CAYM10011436-25
Anti-PSD95 Mouse Monoclonal Antibody [clone: 7E3]
Antibodies
Type: Primary
Antigen: PSD95
Clonality: Monoclonal
Clone: 7E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol PSD95
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 7E3
Reactivity Bovine,Rat,Mouse
Host Mouse
Isotype IgG
Western blot Yes
Immunogen rat recombinant PSD95
Molecular weight 90 kDa
Purification purified by Protein G chromatography
Cross adsorption No
Storage buffer PBS, pH 7,4, containing 0,09% sodium azide and 50% glycerol
Storage temperature –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR