4-Methoxythiobenzamide

Supplier: Cayman Chemical

Synonyms: 4-Methoxybenzothioamide

10552-500 10552-1 10552-5
CAYM10552-500EA 28 EUR
CAYM10552-500 CAYM10552-1 CAYM10552-5
4-Methoxythiobenzamide
4-Methoxythiobenzamide
Formula: C₈H₉NOS
MW: 167.23 g/mol
Boiling Pt: 282
Melting Pt: 155
MDL Number: MFCD00040993
CAS Number: 2362-64-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR