Anti-SMAD8 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
10845-100
CAYM10845-100EA 537 EUR
CAYM10845-100
Anti-SMAD8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMAD8
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR