5-Fluorouracil

Supplier: Cayman Chemical
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

14416-10 14416-25 14416-5 14416-50
CAYM14416-10EA 88 EUR
CAYM14416-10 CAYM14416-25 CAYM14416-5 CAYM14416-50
5-Fluorouracil
5-Fluorouracil
Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347
Melting Pt: 282…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,04208

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR