A potent inhibitor of human SGLT2 that is 1,000-fold less potent at human SGLT1 (Ki values are 2 and 25,000 nM, respectively) 1 * 10 mg
Supplier: Cayman Chemical
A potent inhibitor of human SGLT2 that is 1,000-fold less potent at human SGLT1 (Ki values are 2 and 25,000 nM, respectively) 1 * 10 mg