CCVJ 95

Supplier: Cayman Chemical

Synonyms: 9-(2-Carboxy-2-cyanovinyl)julolidine, (E)-2-Cyano-3-(1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinolin-9-yl)acrylic acid

17870-1 17870-10 17870-5
CAYM17870-1EA 59 EUR
CAYM17870-1 CAYM17870-10 CAYM17870-5
CCVJ 95
CCVJ

CCVJ is a fluorescent molecular rotor characterised by low background fluorescence, low fluorescence anisotropy, and good water solubility. Fluorescent molecular rotors are molecules whose fluorescence is inversely proportional to its intramolecular rotation.


The intramolecular rotation of these probes, and hence their fluorescence, depends on the immediate microenvironment of the probe. As a result, fluorescent molecular rotors are used to evaluate changes in solution and membrane viscosity, polymerisation or aggregation processes, and protein (un)folding.


Moreover, it has large hydrophobic structures, allowing it to associate in a non-covalent manner with hydrophobic pockets in proteins in solution. It is broadly used to monitor changes in solution and molecular characteristics.

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
MDL Number: MFCD00274408
CAS Number: 142978-18-5

Order Now

Specification Test Results

Purity ≥95%
Formal Name 2-cyano-3-(2,3,6,7-tetrahydro-1H,5H-benzo[ij]quinolizin-9-yl)-2-propenoic acid
Formulation A Crystalline Solid
λmax 279, 441 nm
SMILES OC(/C(C#N)=C/C1=CC(CCC2)=C(N2CCC3)C3=C1)=O
InChI Code 1S/C16H16N2O2/c17-10-14(16(19)20)9-11-7-12-3-1-5-18-6-2-4-13(8-11)15(12)18/h7-9H,1-6H2,(H,19,20)/b14-9+
InChI Key JXENNHTVELFRHV-NTEUORMPSA-N
Storage −20 °C
Shipping Room temperature in continental US; may vary elsewhere


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR