(±)-17-HDoHE

Supplier: Cayman Chemical

33650-25 33650-50 33650-100 33650-250
CAYM33650-25EA 89 EUR
CAYM33650-25 CAYM33650-50 CAYM33650-100 CAYM33650-250
(±)-17-HDoHE
(±)-17-HDoHE
Formula: C₂₂H₃₂O₃
CAS Number: 90780-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR