Safety helmets, Concept HeightMaster
Supplier: CENTURION SAFETY PRODUCTS
S08EYFM S08EBFMR S08EWFM S08EKFM S08BFMR S08KFM S08RFMR S08WFM S08WFMR S08YFM S08YFMR
CENUS08EYFMEA 0 EUR
CENUS08EYFM CENUS08EBFMR CENUS08EWFM CENUS08EKFM CENUS08BFMR CENUS08KFM CENUS08RFMR CENUS08WFM CENUS08WFMR CENUS08YFM CENUS08YFMR
Safety helmets, Concept HeightMaster
Head Protection Safety Helmets
Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene). The Concept HeightMaster has been designed for applications where the wearer is working at height, where the suspension system and the helmet design and structure will reduce the risk of the helmet coming off as a result of an impact or a fall.
  • With extended nape area
  • Six point terylene inner cradle
  • Highly absorbent hydro-flock sweatband
  • With increased side rigidity (additional lateral deformity test)
  • Ventilation through wide ventilation slots
  • With or