Face shields, chin guard system, Connect®
Supplier: CENTURION SAFETY PRODUCTS
S910 S911 S912
CENUS910EA 21.86 EUR
CENUS910 111-1580 CENUS912
Face shields, chin guard system, Connect®
Face Shields
These clear face shields are available with acetate or PC screens.
  • Chinguard screens have holes at bottom in order to attach chinguard
  • Can be used in combination with a browguard when a safety helmet is not required
Certifikace: EN 166 B