2,4,6-Tribromoanisole

Supplier: CUSTOM MADE CHEMICALS LAB

Synonyms: 1,3,5-Tribromo-2-methoxybenzene

SB19790.100MG
CMPLSB19790.100MGEA 0 EUR
CMPLSB19790.100MG
2,4,6-Tribromoanisole
2,4,6-Tribromoanisole
Formula: C₇H₅Br₃O
MW: 344.83 g/mol
Boiling Pt: 298 °C
Melting Pt: 88 °C
Density: 25 °C
MDL Number: MFCD00192510
CAS Number: 607-99-8

Order Now

Specification Test Results

XLogP3 4.1
Hydrogen bond donor count 0
Rotatable bond count 0
Exact mass 1
Monoisotopic mass 343.78700 g/mol
Topological polar surface area 341.78905 g/mol
Heavy atom count 9.2 Ų
Formal charge 11
Complexity 0
Isotope atom count 119
Defined atom stereocenter count 0
Undefined atom stereocenter count 0
Defined bond stereocenter count 0
Undefined bond stereocenter count 0
Covalently-bonded unit count 1
Compound is canonicalized Yes
Hydrogen bond acceptor count 1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR