Cleanroom wipes, Anticon® Gold HeavyWeight™
Supplier: CONTEC
492224-789 492224-790 492224-783 492224-798
115-0435EA 121 EUR
115-0435 115-0436 CONT492224-783 CONT492224-798
Cleanroom wipes, Anticon® Gold HeavyWeight™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
100% no-run interlock knit polyester wipes with sealed edges and increased sorbency.
  • Sealed edges to reduce particle and fibre generation
  • High sorptive capacity and fast adsorption rate for superior spill control
  • High tensile strength to eliminate tearing and snagging in tough cleaning applications
  • Patented textile technology achieves superior sorption characteristics for critical applications
  • Specially processed for superior cleanliness and sorbency
Anticon® Gold HeavyWeight™ cleanroom wipes are made from 100% continuous filament polyester with aMore Product Information